Anti-APOBEC3C Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310103EA 381 CHF
ORIGTA310103
Anti-APOBEC3C Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: APOBEC3C (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR