Anti-ADCY6 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310097EA 448 CHF
ORIGTA310097
Anti-ADCY6 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ADCY6 (adenylate cyclase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR