Anti-ADAMTS2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA310092EA 448 CHF
ORIGTA310092
Anti-ADAMTS2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ADAMTS2 (ADAM metallopeptidase with thrombospondin type 1 motif, 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR