Anti-AAMP Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310084EA 448 CHF
ORIGTA310084
Anti-AAMP Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:250

Type: Primary
Antigen: AAMP (angio-associated, migratory cell protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR