Anti-PLXNB1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310077EA 418 CHF
ORIGTA310077
Anti-PLXNB1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: PLXNB1 (plexin B1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR