Anti-CDH15 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310065EA 418 CHF
ORIGTA310065
Anti-CDH15 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CDH15 (cadherin 15, type 1, M-cadherin (myotubule))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR