Anti-LTB4R Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310062EA 381 CHF
ORIGTA310062
Anti-LTB4R Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: LTB4R (leukotriene B4 receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR