Anti-CLOCK Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA310045EA 448 CHF
ORIGTA310045
Anti-CLOCK Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CLOCK (clock circadian regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR