Anti-ABCD4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310031EA 373 CHF
ORIGTA310031
Anti-ABCD4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ABCD4 (ATP-binding cassette, sub-family D (ALD), member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR