Anti-ABCD3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA310030EA 381 CHF
ORIGTA310030
Anti-ABCD3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ABCD3 (ATP-binding cassette, sub-family D (ALD), member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR