Anti-TPCN2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310027EA 418 CHF
ORIGTA310027
Anti-TPCN2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:20000-1:80000; Immunochemistry: 1:100-1:500

Type: Primary
Antigen: TPCN2 (two pore segment channel 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR