Anti-TACSTD2 Mouse Monoclonal Antibody

Supplier: OriGene
ORIGTA310025EA 660 CHF
ORIGTA310025
Anti-TACSTD2 Mouse Monoclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: TACSTD2 (tumor-associated calcium signal transducer 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR