Anti-APP Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310001EA 668 CHF
ORIGTA310001
Anti-APP Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR