Anti-PRKDC Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309996EA 668 CHF
ORIGTA309996
Anti-PRKDC Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: PRKDC (protein kinase, DNA-activated, catalytic polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR