Anti-CDC25A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309985EA 645 CHF
ORIGTA309985
Anti-CDC25A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR