Anti-IGF1R Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309975EA 717 CHF
ORIGTA309975
Anti-IGF1R Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR