Anti-G3BP1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309970EA 717 CHF
ORIGTA309970
Anti-G3BP1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: G3BP1 (GTPase activating protein (SH3 domain) binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR