Anti-STAT6 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309960EA 668 CHF
ORIGTA309960
Anti-STAT6 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: STAT6 (signal transducer and activator of transcription 6, interleukin-4 induced)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR