Anti-STAT5A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309958EA 702 CHF
ORIGTA309958
Anti-STAT5A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR