Anti-ELK1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309950EA 717 CHF
ORIGTA309950
Anti-ELK1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: ELK1 (ELK1, member of ETS oncogene family)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR