Anti-GNRHR Mouse Monoclonal Antibody [clone: AT2.G7]
Supplier: OriGene
ORIGTA309941EA 660 CHF
ORIGTA309941
Anti-GNRHR Mouse Monoclonal Antibody [clone: AT2.G7]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:200
Type: Primary
Antigen: GNRHR (gonadotropin-releasing hormone receptor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human