Anti-CTHRC1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309936EA 652 CHF
ORIGTA309936
Anti-CTHRC1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; ELISA: 1:2000-5000

Type: Primary
Antigen: CTHRC1 (collagen triple helix repeat containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR