Anti-ZMPSTE24 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309934EA 777 CHF
ORIGTA309934
Anti-ZMPSTE24 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1.0-2.0 μg/ml

Type: Primary
Antigen: ZMPSTE24 (zinc metallopeptidase STE24)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR