Anti-TP53INP2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309919EA 740 CHF
ORIGTA309919
Anti-TP53INP2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: TP53INP2 (tumor protein p53 inducible nuclear protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR