Anti-TNC Mouse Monoclonal Antibody [clone: 4C8MS]
Lieferant: OriGene
ORIGTA309913EA 841 CHF
ORIGTA309913
Anti-TNC Mouse Monoclonal Antibody [clone: 4C8MS]
Antikörper
Recommended Dilutions:
Type: Primary
Antigen: TNC (tenascin C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat