Anti-STK33 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309885EA 755 CHF
ORIGTA309885
Anti-STK33 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: STK33 (serine/threonine kinase 33)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR