Anti-SPTBN2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309879EA 841 CHF
ORIGTA309879
Anti-SPTBN2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: SPTBN2 (spectrin, beta, non-erythrocytic 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR