Anti-SPHK1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309876EA 672 CHF
ORIGTA309876
Anti-SPHK1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: SPHK1 (sphingosine kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR