Anti-SLC40A1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309869EA 904 CHF
ORIGTA309869
Anti-SLC40A1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: SLC40A1 (solute carrier family 40 (iron-regulated transporter), member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR