Anti-SLC18A2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309861EA 1010 CHF
ORIGTA309861
Anti-SLC18A2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: SLC18A2 (solute carrier family 18 (vesicular monoamine transporter), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR