Anti-SGOL1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309856EA 755 CHF
ORIGTA309856
Anti-SGOL1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: SGOL1 (shugoshin-like 1 (S. pombe))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR