Anti-SFRP5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309855EA 672 CHF
ORIGTA309855
Anti-SFRP5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: SFRP5 (secreted frizzled-related protein 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR