Anti-SERF1A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309849EA 740 CHF
ORIGTA309849
Anti-SERF1A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: SERF1A (small EDRK-rich factor 1A (telomeric))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR