Anti-SEMA3B Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309848EA 841 CHF
ORIGTA309848
Anti-SEMA3B Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: SEMA3B (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR