Anti-RPS27A Mouse Monoclonal Antibody [clone: Ubi-1]
Supplier: OriGene
ORIGTA309840EA 929 CHF
ORIGTA309840
Anti-RPS27A Mouse Monoclonal Antibody [clone: Ubi-1]
Antibodies
Recommended Dilutions:
Type: Primary
Antigen: RPS27A (ribosomal protein S27a)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now