Anti-RPE65 Mouse Monoclonal Antibody [clone: 401.8B11.3D9]
Lieferant: OriGene
ORIGTA309839EA 904 CHF
ORIGTA309839
Anti-RPE65 Mouse Monoclonal Antibody [clone: 401.8B11.3D9]
Antikörper
Recommended Dilutions:
Type: Primary
Antigen: RPE65 (retinal pigment epithelium-specific protein 65kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now