Anti-RBFOX3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309832EA 841 CHF
ORIGTA309832
Anti-RBFOX3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: RBFOX3 (RNA binding protein, fox-1 homolog (C. elegans) 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR