Anti-RASSF5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309829EA 633 CHF
ORIGTA309829
Anti-RASSF5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: RASSF5 (Ras association (RalGDS/AF-6) domain family member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR