Anti-PRMT2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309820EA 686 CHF
ORIGTA309820
Anti-PRMT2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: PRMT2 (protein arginine methyltransferase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR