Anti-POLR2A Mouse Monoclonal Antibody [clone: 4H8]
Supplier: OriGene
ORIGTA309814EA 777 CHF
ORIGTA309814
Anti-POLR2A Mouse Monoclonal Antibody [clone: 4H8]
Antibodies
Recommended Dilutions:
Type: Primary
Antigen: POLR2A (polymerase (RNA) II (DNA directed) polypeptide A, 220kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now