Anti-PLIN1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309806EA 777 CHF
ORIGTA309806
Anti-PLIN1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: PLIN1 (perilipin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR