Anti-PKM Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309804EA 777 CHF
ORIGTA309804
Anti-PKM Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR