Anti-PDIA2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309796EA 777 CHF
ORIGTA309796
Anti-PDIA2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: PDIA2 (protein disulfide isomerase family A, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR