Anti-P2RY2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309786EA 777 CHF
ORIGTA309786
Anti-P2RY2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: P2RY2 (purinergic receptor P2Y, G-protein coupled, 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR