Anti-OGG1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA309779EA 904 CHF
ORIGTA309779
Anti-OGG1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: OGG1 (8-oxoguanine DNA glycosylase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR