Anti-NES Mouse Monoclonal Antibody [clone: 10C2]

Supplier: OriGene
ORIGTA309763EA 761 CHF
ORIGTA309763
Anti-NES Mouse Monoclonal Antibody [clone: 10C2]
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: NES (nestin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR