Anti-NES Mouse Monoclonal Antibody [clone: 4D11]
Supplier: OriGene
ORIGTA309762EA 761 CHF
ORIGTA309762
Anti-NES Mouse Monoclonal Antibody [clone: 4D11]
Antibodies
Recommended Dilutions:
Type: Primary
Antigen: NES (nestin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat