Anti-MAP1B Rabbit Polyclonal Antibody [clone: SuperBUGS]
Supplier: OriGene
ORIGTA309739EA 966 CHF
ORIGTA309739
Anti-MAP1B Rabbit Polyclonal Antibody [clone: SuperBUGS]
Antibodies
Recommended Dilutions:
Type: Primary
Antigen: MAP1B (microtubule-associated protein 1B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now