Anti-LMNA Mouse Monoclonal Antibody [clone: 5H8-B4]
Lieferant: OriGene
ORIGTA309731EA 740 CHF
ORIGTA309731
Anti-LMNA Mouse Monoclonal Antibody [clone: 5H8-B4]
Antikörper
Recommended Dilutions:
Type: Primary
Antigen: LMNA (lamin A/C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human