Anti-KLK5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA309721EA 633 CHF
ORIGTA309721
Anti-KLK5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: KLK5 (kallikrein-related peptidase 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR